missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Antimicrobial Polypropylene Barbed Fittings

These single barb fittings offer natural silver protection for the fitting in applications where microorganism growth is likely. Using the AgION™ technology the fittings inhibit the growth of microorganisms on the surface of the fitting.

Brand:  Masterflex™ 06368-04

 View more versions of this product

Product Code. 11700339

  • 26.30 € / Pack of 10
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prevents the growth of bacteria, fungi, mold and mildew
  • Meets EPA standards
  • NSF 51, 61 and FDA compliant
  • Silver protection is not temperature sensitive and remains effective over time
  • Silver treatment will not contaminate your sample
  • Slip flexible tubing or hose onto barb to create a seal that typically holds pressures up to 30 psi (2 bar)
  • Manufactured from polypropylene
  • Sterilization method of ethylene oxide (ETO) or gamma
  • Available in straight and Y shape configurations
Specifications

Specifications

Barbed Fitting
1/4 in. tubing I.D.
Microbial growth
Ethylene Oxide, Gamma
Straight
30 psi (2 bar)
Polypropylene
10 Pack
3.56mm orifice
8 mm
Light Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.