missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Polypropylene Reducing Hose Connectors
Use Buerkle™ Polypropylene Reducing Hose Connectors to make a connections between larger and smaller hoses.
Brand: Buerkle™ 8703-0412
20.05 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polypropylene
- Sterilizable
Specifications
EU Foodstuffs, RoHS | |
Autoclavable | |
4 to 8/12 to 16 mm Hose |
Reducing Hose Connector | |
Polypropylene | |
10/Pk.; 10/Scale |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction