Learn More
Cole-Parmer™ Polypropylene Copolymer Lab Containers
Brand:  Cole-Parmer™ 06727-35
Product Code. 15237299
  • 258.00 € / Pack of 50
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
50 Pk.
Translucent
PPCO
152.4 mm
2000 mL
Lab Container
Single-use