Learn More
Brand™ Polypropylene Wide Neck Erlenmeyer Flasks
Flasks with high clarity for visibility of contents during routine laboratory procedures.
Brand:  Brand™ 93172
 View more versions of this product
Product Code. 10799581
  • 40.30 € / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Provided with screw cap closure
  • With blue graduation marks
  • Neck with standard ground socket
  • Provided in packs of 6, except for 1000mL flask, which comes in a pack of 4
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
110 mm
6
Screw Cap
Polypropylene
34/35
GL 40
64 mm