Learn More
Masterflex™ Polypropylene Female Luer Bulkhead Fitting
Brand:  Masterflex™ 45508-30
 View more versions of this product
Product Code. 15253265
  • 30.70 € / Pack of 25
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Female Luer Bulkhead Fitting
Fits 1/4-28 UNF thread to 1/16 in. hose barb
25 Pack
Polypropylene
Ideal for a variety of applications, including syringes or small tubing