missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Funnel Holders

Secure funnel holders onto rod with included olives. Kartell™ Polypropylene Funnel Holders are adaptable to a variety of types and sizes of funnels.

Brand:  Kartell™ 0026900

Product Code. 10386201

  • 21.65 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Double holder for funnels as small as 25 mm (with supplied adaptor) up to 170 mm
  • Use with powder funnels with stem diameter not exceeding 35 mm when adaptor is removed
  • Two olives provided for use with rods from 8 to 14 mm
Specifications

Specifications

Polypropylene
25 to 170mm funnels; powder funnels
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.