Learn More
Sharpsguard™ dani 22 Multi-Purpose Waste Container
Ideal for the safe collection and transportation of glass and crockery. Sharpsguard™ dani 22 Multi-Purpose Waste Container is color-coded with a solid orange lid, handle and label, making it easy to distinguish from other waste containers.
Brand:  Daniels Healthcare DD820
Product Code. 12322449
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Should be disposed of in landfill sites and therefore is not suitable for contaminated items
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications
Specifications
21.01 L
Orange
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sharpsguard™ dani 22 Multi-Purpose Waste Container > Capacity: 22L