missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Huenersdorf™ Polypropylene Graduated Beaker with Closed Handle and 2 Scales
Huenersdorf™ Polypropylene Graduated Beaker with Closed Handle and 2 Scales have both injection-molded and blue-embossed scales. Graduations are marked in capacity-appropriate increments.
Specifications
Color | Transparent |
---|---|
Material | Polypropylene |
For Use With (Application) | For Use in The Laboratory, Workshop and Household |
Includes | With Injection Molded Scale and Blue Embossed Scale |
Quantity | 1 Ea. |
Description
- Polypropylene beaker
- Graduated
- Blue embossed scale
- Injection molded scale
- Transparent
Specifications
Transparent | |
For Use in The Laboratory, Workshop and Household | |
1 Ea. | |
Autoclavable |
Polypropylene | |
With Injection Molded Scale and Blue Embossed Scale | |
Beaker | |
Yes |