Learn More
Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes
Use these tubes for a variety of applications. Fisherbrand™ Polypropylene Graduated Microcentrifuge Tubes are ideal for use with PCR systems and centrifugation.
21.70 €
Specifications
Autoclavable | Autoclavable |
---|---|
Capacity (Metric) | 1.5 mL |
Material | Polypropylene |
For Use With (Application) | For PCR, Centrifugation |
No. per Pack | 500/Pk. |
Description
- Polypropylene graduated tubes
- Autoclavable
- Available in a variety of colors for easy identification
- Available with pick-up tab
Specifications
Autoclavable | |
Polypropylene | |
500/Pk. | |
Snap Cap |
1.5 mL | |
For PCR, Centrifugation | |
Yes |