Learn More
Eppendorf™ Polypropylene Graduated Microtubes
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
Brand: Eppendorf™ 0030121570
33.21 EUR valid until 2024-12-31
Use promo code "21938" to get your promotional price.
Description
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. EppendorfTM Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
- Biohazardous substances can be removed, free of aerosols, through a thin membrane in the center of the lid with a cannula
- Lid easily opens with one hand
- These tubes for 0.5mL, 1.5mL and 2mL have a graduation scale and writing surfaces
- The 0.5mL tube is optimized for use in PCR* experiments
- Assorted color packs are a mixture of yellow, red, blue, green and colorless
- Autoclavable micropestle for re-suspending pellets
- Can be centrifuged at up to 23,500xgr
Specifications
30,000 x g | |
0.5 mL | |
Polypropylene | |
50/Pk. | |
Biopur | |
Hinged Snap Cap |
Autoclavable | |
Clear | |
PCR, Centrifugation | |
Sterile | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.