Learn More
Buerkle™ Polypropylene Straight Connectors with Conical Nozzles
Manufactured from polypropylene for high thermal resistance. Buerkle™ Polypropylene Straight Connectors with Conical Nozzles resist temperatures up to 135°C.
Brand:  Buerkle™ 8700-0810
 View more versions of this product
Product Code. 10198251
  • 22.65 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Cylindrical straight hose connector with conical nozzles
  • Polypropylene
  • Autoclavable to 121°C
  • Suitable for foodstuff
  • Available in a wide range of diameters for varying hose sizes
Specifications
Specifications
EU Foodstuffs, RoHS
Autoclavable
7 to 10 mm Hose
Straight
Hose Tubing Connector
Polypropylene
10/Pk.; 10/Scale