missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Straight Connectors with Conical Nozzles

Manufactured from polypropylene for high thermal resistance. Buerkle™ Polypropylene Straight Connectors with Conical Nozzles resist temperatures up to 135°C.

Brand:  Buerkle™ 8700-1416

 View more versions of this product

Product Code. 10486891

  • 21.85 € / Pack of 10
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cylindrical straight hose connector with conical nozzles
  • Polypropylene
  • Autoclavable to 121°C
  • Suitable for foodstuff
  • Available in a wide range of diameters for varying hose sizes
Specifications

Specifications

EU Foodstuffs, RoHS
Autoclavable
13 to 15 mm Hose
Straight
Hose Tubing Connector
Polypropylene
10/Pk.; 10/Scale
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.