Learn More
Brady™ Polypropylene: ISO Safety Sign - Collection of fire-fighting equipment
Sign compliant to ISO 7010.
Brand:  Brady™ 817363
 View more versions of this product
Product Code. 15644397
  • 6.75 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
Polypropylene
Indoor and Outdoor
1.501 mm
-40°C to 120°C
Global
PIC F004-100x100-PP-CRD/1
1 Ea.
100 mm
100 mm
Square
White on Red