Learn More
Brady™ Polypropylene: ISO Safety Sign - Fire ladder
Sign compliant to ISO 7010.
Brand:  Brady™ 817207
 View more versions of this product
Product Code. 15634387
  • 16.10 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
Polypropylene
Indoor and Outdoor
1.501 mm
-40°C to 120°C
Global
PIC F003-250x250-PP-CRD/1
1 Ea.
250 mm
250 mm
Square
White on Red