missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Polypropylene: ISO Safety Sign - Use this walkway

Sign compliant to ISO 7010.

Brand:  Brady™ 821406

 View more versions of this product

Product Code. 15604567

  • 37.00 € / Each
Estimated Shipment: 16-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene
  • For indoor and outdoor use
Specifications

Specifications

ISO 7010
1 Ea.
Indoor and Outdoor
-40°C to 120°C
Global
Polypropylene
315 mm
1.501 mm
Round
White on Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers