missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polypropylene Labels for BradyJet J4000
This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.
Brand: Brady™ 152779
This item is not returnable.
View return policy
Description
- Matte white polypropylene with acrylic adhesive is Inkjet printable
- Low-cost, full-color GHS labeling option withstands chemical exposure
- Service temperature of -80°C to 70°C
- An ink cartridge is required to print this material
Specifications
Autoclavable | |
Permanent Acrylic | |
Matte | |
Ink Jet | |
Hazard Communication Identification, Equipment Labeling, Laboratory Labeling, General Identification | |
1 | |
Rectangle | |
0.13 mm | |
104.9 mm | |
155 mm |
J40-262-7425J | |
White | |
Polypropylene (Inkjet-receptive) | |
Non-sterile | |
BradyJet J4000 Color Label Printer | |
200 Labels | |
-80°C to +70°C | |
161.9 mm | |
104.9 mm | |
155 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction