Learn More
Cole-Parmer™ Elbow Adapter, Compression x NPT(M), Polypropylene
Cole-Parmer™ Elbow Adapter, Compression x NPT(M), Polypropylene features thread and a compression connections for a more permanent elbow adapter fitting.
Brand:  Cole-Parmer™ 06444-20
Product Code. 11810110
  • 122.00 € / Pack of 25
Estimated Shipment: 10-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sturdy threaded fittings are designed for more permanent installation.
  • Use with our rigid tubing for high-pressure applications.
Specifications
Specifications
Male Pipe Elbow Adapter
Polypropylene
25 Pack
30 mm
Autoclavable
1/4 in. OD tubing, 1/8 in. NPT connection
Autoclave Only
Black