Learn More
Cytiva MBS II Microbiological Filtration System Filter Units
Eliminates time-intensive sterilizing procedures. Whatman™ MBS II Microbiological Filtration System Filter Units combine the membrane and funnel into one pre-assembled unit for simple, high-throughput filtration with reproducible results and minimal risk of cross-contamination.
Brand: Cytiva 10445903
Description
The Whatman™ MBS II Microbiological Filtration System was designed to provide complete control over the sample filtration process. A pre-assembled all-in-one filtration unit is attached to a vacuum manifold (sold separately). The sample is poured into the filter unit for sterile filtration. Then, the unit is detached and the membrane is just removed and transferred to an agar plate. A new sterile filtration unit is used for the next sample, with no sterilization of the system required.
- Easy handling: Ready-to-use units eliminate complicated sterilization procedures
- Reduced testing time: No need to sterilize between samples, reducing testing time by 50%
- Compatible: Membrane is compatible with all agar and Petri dish formats
- No flaming required: Combined filtration unit minimizes the risk of cross-contamination
- Sterile venting: Sample is protected by a vented lid during filtration
- Reproducible results: Closed system filtration unit reduces the chance of external error
- Applications: Pharmaceutical manufacturing, Processed water tests, Bioburden tests on raw materials, Quality control of non-sterile end products, Drinking water evaluations
The mixed cellulose ester membrane (ME) is hydrophillic and is ideal for aqueous solutions. The white filter features a black grid for easy quantitative evaluation of microorganism counts. After filtration, the membrane is removed from the filter unit and incubated on a nutrient medium so the individual colonies can be evaluated.
Specifications
White filter with black grd | |
Polypropylene | |
Sterile filtration of microbiological samples, e.g. pharmaceutical testing | |
24Pack |
Vacuum manifold (mfr. no. 10445990) | |
0.2μm | |
Membrane: Mixed cellulose esters (ME) | |
100 mL |