missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Microcentrifuge Tube

Screw cap lid sold separately.

Brand:  BRAND™ 780762

104.65 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10413675

  • 138.00 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene (PP) construction
  • Sterile
  • Operating temperature: -196 to 121°C
  • Autoclavable
  • In accordance with DIN EN 285
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.5 mL
44 mm
1000
20.000 G
Polypropylene
Without base
–196 to 121°C
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.