missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Microcentrifuge Tubes

Use these tubes for high speed centrifugation applications. Fisherbrand™ Polypropylene Microcentrifuge Tubes feature a frosted writing surface on closure and side of tube (except for natural tubes) which allows for easy and convenient sample identification.

Brand:  Fisherbrand™ FB74023

24.83 EUR valid until 2024-12-31
Use promo code "21938" to get your promotional price.


 View more versions of this product

Product Code. 11508232

  • 38.45 € / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Graduated polypropylene tubes
  • Suitable for withstanding the stress of high speed centrifugation up to 20,000xg
  • One-piece construction incorporates a snug fitting and reliable cap and the bottom is reinforced for added protection against leakage
  • Autoclavable
Specifications

Specifications

20,000 x g
Autoclavable
Natural
Microcentrifuge
1000/Pk.
Polypropylene
0.5 mL
Polypropylene
1000/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers