missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Microplates, 384-well
Offers higher well density for economical use of expensive reagents. Fisherbrand™ Polypropylene Microplates, 384-well include round-shaped wells for optimal sample recovery.
429.00 € - 526.00 €
Specifications
Certifications/Compliance | ANSI™ standard footprint |
---|---|
Format | Standard |
Array | 16 x 24 |
Material | Polypropylene |
No. of Wells | 384 |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11977954
|
Fisherbrand™
236200 |
Yellow |
429.00 €
Pack of 120 |
In Stock
Log in to see stock. |
|||||
11987954
|
Fisherbrand™
236300 |
Blue |
526.00 €
Pack of 120 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
|||||
Description
- Ideal for storage of compounds and nucleic acid libraries
- Chemical resistant
- Withstands temperatures from -80° to +80°C
- Conform to ANSI™ standard footprint for optimal use in automated handling and liquid-dispensing systems
Specifications
ANSI™ standard footprint | |
16 x 24 | |
384 | |
120/Pk. |
Standard | |
Polypropylene | |
Round | |
-80°C to +80°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title