Learn More
Brady™ Polypropylene and Molded Glass-filled Nylon No Hole Circuit Breaker Lockout
These No-Hole breaker lockouts are extremely versatile and easy to install using patented thumbscrew design - no screwdrivers needed. Simply tighten lockout securely onto switch tongue, pull cover over thumbscrew and lock cover to prevent clamp from being loosened.
Brand:  Brady™ 065397
 View more versions of this product
Product Code. 11701895
  • 17.50 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Detachable Cleats (included) expand the range of applicable breakers.
  • Accepts lock shackles up to 7 mm in diameter.
Specifications
Specifications
Circuit Breaker Lockout
Glass-filled Nylon
No Hole Circuit Breaker Lockout - 480/600 V Device
7 mm
82.60 x 40.30 mm
1.588 in.
1 Ea.
82.63 mm
Red
OSHA 29 CFR 1910.147
0.3 in.
55.25 x 82.63 x 40.34 mm
Lockout Tagout
40.34 mm
3.253 in.