Learn More
Brady™ Polypropylene and Molded Glass-filled Nylon No Hole Circuit Breaker Lockout
These No-Hole breaker lockouts are extremely versatile and easy to install using patented thumbscrew design - no screwdrivers needed. Simply tighten lockout securely onto switch tongue, pull cover over thumbscrew and lock cover to prevent clamp from being loosened.
Brand:  Brady™ 065396
 View more versions of this product
Product Code. 11791885
  • 15.75 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Detachable Cleats (included) expand the range of applicable breakers.
  • Accepts lock shackles up to 7 mm in diameter.
Specifications
Specifications
Circuit Breaker Lockout
Glass-filled Nylon
No Hole Circuit Breaker Lockout - 120/277 V Device
7 mm
56 x 24.10 mm
0.95 in.
1 Ea.
56.01 mm
Red
OSHA 1910.147
0.3 in.
25.40 x 56.01 x 24.13 mm
Lockout Tagout
24.13 mm
2.205 in.