Learn More
Ratiolab™ Polypropylene Multi Purpose Cup
Product Code. 15303492
Click to view available options
Capacity (Metric):
60 mL
Unit Size:
Pack of 500
Product Code. 15303492
Brand: Ratiolab™ 5921176
Only null left
Add to basket
Edge
Add to basket
Specifications
Sterility Sterile
Quantity 500 Pk.
Capacity (Metric) 60 mL
Product Type Multipurpose
Material Polypropylene
For Use With (Application) For sampling, transport and storage of solid, pasty and liquid samples
For Use With (Equipment) For sampling, transport and storage of solid, pasty and liquid samples