Learn More
Fisherbrand™ Polypropylene Natural Pipet Tip
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Natural Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
Brand:  Fisherbrand™ FB34631
 View more versions of this product
Product Code. 11597452
  • 19.90 € / Pack of 1000
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Specifications
250 to 1000 μL
Natural
Autoclavable
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Polypropylene Natural Pipet Tip > Capacity: 250 to 1000uL