Learn More
Fisherbrand™ Polypropylene Pipet Tip G
Make pipetting much simpler and safe for even the most sensitive applications. Fisherbrand™ Polypropylene Pipet Tips G are perfectly fit for manual or electronic, single or multichannel pipets.
Brand:  Fisherbrand™ FB64006
 View more versions of this product
Product Code. 12399365
  • 20.05 € / Pack of 192
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of high quality polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to the stable shape
  • Autoclavable
  • Guaranteed dust-free due to special ionization treatment
  • Available pre-sterilized or non-sterile
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
  • Aeroject™ tips with special filters made of hydrophobic PE-HD as
  • Aerosol barrier and for protection against contamination
  • Suitable for all commonly used pipet types
Specifications
Specifications
0.5 to 10 μL
Autoclavable
MultiTray
Crystal
Rack
Polypropylene