Learn More
VITLAB™ Polypropylene Pipette Stand
Manufactured from polypropylene. VITLAB™ Polypropylene Pipette Stand has a rotatable, ribbed base plate in which the pipette tips can be gently seated.
Brand: VITLAB™ 79194
This item is not returnable.
View return policy
Description
- Polypropylene
- 94 bore holes of different diameters
- Easy assembly
Specifications
Pipette Stand | |
94 Any Size | |
470 mm |
Polypropylene | |
1 Each | |
230 mm |
Product Content Correction
Product Title
VITLAB™ Polypropylene Pipette Stand > Holds: 94 Any Size
Spot an opportunity for improvement?Share a Content Correction