missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Plain Test Tube

Supplied with polyethylene quick closure leak-proof screwcaps. Kartell™ Polypropylene test tubes are suitable for centrifugation.

Brand:  Kartell™ 8830303

Product Code. 11944279

  • 172.00 € / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from polypropylene for optimum chemical and thermal stability
  • Leak-proof, and suitable for centrifugation
  • The design of the cap avoids any contamination due to drops of liquid on the inner surface
  • Maximum force: 4000 x g
  • Available in: blue (04) - red (10)- yellow (06) - white (03)
  • CE-IVD approved
Specifications

Specifications

CE and IVD
Polyethylene
15 mL
Single-use
1000 Pack
Autoclavable
Screw Cap
White
Test tube
Polypropylene
Suitable for centrifugation
Non-sterile
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.