missing translation for 'onlineSavingsMsg'
Learn More
3M™ White Disposable Protective Coveralls, 4515 Series Product Code.: 12912275
Estimated Shipment: 16-05-2025
to see stock.
Add to basket

3M™ White Disposable Protective Coveralls, 4515 Series

Product Code. 12912275
Large, 69 to 71 in., 174 to 181 cm
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
194 to 200 cm
‎186 to 194 cm
‎200 to 206 cm
This item is not returnable. View return policy

Product Code. 12912275

Brand: 3M™ 7000089613

Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Provides PPE Directive Category III, Type 5/6 protection. The Protective Coverall 4515 Series protect from hazardous dusts such as asbestos fibers.

  • Breathable material reduces heat build-up and discomfort
  • Excellent barrier against dry particles and limited liquid splash (CE Type 5/6)
  • Robust, highly breathable 50gsm SMS material
  • Meets inward leakage requirements of EN 1073-2 for protection against nuclear particles
  • Two-way zipper with fully sealable storm flap
  • Two-panel hood design
  • Elasticated wrists, waist, and ankles for increased comfort and freedom of movement
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size Large
Gender Unisex
Material SMS Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class PPE Directive Category III, Type 5/6
Model 4515W
Size (English) Chest 39 to 43 in.
Size (Metric) Chest 100 to 108 cm
Height (English) 69 to 71 in.
Height (Metric) 174 to 181 cm
Show More Show Less
  • 3M Safety Solutions - Protective coveralls overview

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ White Disposable Protective Coveralls, 4515 Series >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.