Learn More
Polypropylene Ratchet Clips
Use these clips for a variety of laboratory applications. S Murray™ Polypropylene Ratchet Clips are suitable for use with flexible tubing up to 10mm diameter.
Brand: S Murray SAMCO L140/02
Description
- Polypropylene
- Pack of 10
Specifications
Polypropylene | |
Suitable for use with flexible tubing up to 10mm diameter | |
Ratchet Clip |
Suitable for use with flexible tubing up to 10mm dia. | |
10 Pack |
Product Content Correction
Product Title
Polypropylene Ratchet Clips > Fits Tubing: Up to 10mm diameter
Spot an opportunity for improvement?Share a Content Correction