Learn More
Fisherbrand™ PPCO Rectangular Carboy with Spigot
For Use With: Ideal for storing and dispensing lab water and reagents; Autoclavable; Closure Material: PPCO; Color: White
134.00 € - 191.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | FDA, USP VI |
Closure Material | PPCO |
Color | White |
For Use With (Application) | Ideal for storing and dispensing lab water and reagents |
Description
Value your lab can rely on. Fisherbrand PPCO Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Ideal for storing and dispensing lab water and reagents
- Durable construction for long service life
- Molded graduations for 1 gallon and 51 increments
- Choice of materials for general use or autoclavability
- Space-saving rectangular design
- Wide mouth closure allows for easy filling and cleaning access
- Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
- Spigot spout without the adapter fits 5/8-in. ID tubing
- Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
Specifications
Autoclavable | |
PPCO | |
Ideal for storing and dispensing lab water and reagents | |
Wide | |
Rectangular |
FDA, USP VI | |
White | |
PPCO | |
Carboy with Spigot |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.