Learn More
Bel-Art™ Scienceware™ Polypropylene Scoop
Designed for retrieving samples from a larger source. Bel Art™ Polypropylene Scoop is excellent for scooping anything from bulk powder to ice.
Brand:  Bel-Art™ Scienceware™ F36758-0000
 View more versions of this product
Product Code. 11701495
  • 142.00 € / Pack of 6
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rounded corners for easy cleaning
  • Made from rigid polypropylene
  • Looped handle for hanging 
  • Holds approximately 2 liters (one half gallon)
Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes. Stated volumes represent typical heaping samples.
Specifications
Specifications
Scoop
2 L
349 x 148 x 89 mm
349 mm
122 mm
Polypropylene
349 x 148 mm
89 mm
227 mm
148 mm