Learn More
Cytiva Polypropylene Screw Lid GL45 Kit
Used to secure containers. Cytiva Polypropylene Screw lid GL45 Kit comes with screw lid and cap membrane.
Brand: Cytiva 11-0004-10
This item is not returnable.
View return policy
Description
- The kit contains 1 x Screw lid 1 x Cap membrane
- The membrane should not be used when working with organic solvent.
- Lid: Polypropylene (PP)
- Membrane: Ethylenepropylenediene (EPDM)
Specifications
Lid | |
Screw Lid GL45 Kit | |
1 Kit |
Polypropylene, Ethylenepropylenediene | |
Screw lid, Cap membrane |