missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Polypropylene Stackable Beaker

Use these stackable beakers for a variety of general lab applications. VITLAB™ Polypropylene Stackable Beakers feature a stable handle and easily readable, printed black scale on both sides. Volume is equally visible for left and right handers.

Brand:  Vitlab™ 483941

 View more versions of this product

Product Code. 10261232

  • 131.00 € / Pack of 12
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Highly transparent
  • To preserve markings, cleaning at no higher than 60°C is recommended
  • Suitable for foodstuffs
Specifications

Specifications

148 mm
2000 mL
Polypropylene
212 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.