missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Storage Box with lid, Autoclavable
Temperature resistant from -80° up to +100°.
Brand: Fisherbrand™ 12 21 3141
This item is not returnable.
View return policy
Description
- For 1.5mL, 1.8mL, 2mL vials or 2mL shell vials
- Includes cover
- Dimension (L x W x H): 67 x 67 x 45mm
- 16 cavities
- Stackable for efficient storage
Specifications
Transparent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction