Learn More
BRAND™ Polypropylene Thread Adapters
These Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use and are ideal for use with Dispensetter bottletop dispensers.
9.65 € - 42.75 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
10480223
|
BRAND™
704424 |
For ground joint NS 24/29 |
17.85 €
Each |
In Stock
Log in to see stock. |
||||||
10471183
|
BRAND™
704322 |
For bottle thread GL 22 |
10.40 €
Each |
In Stock
Log in to see stock. |
||||||
10212371
|
BRAND™
704325 |
For bottle thread GL 24-25 |
9.65 €
Each |
In Stock
Log in to see stock. |
||||||
10084570
|
BRAND™
704330 |
For bottle thread GL 30 |
12.60 €
Each |
In Stock
Log in to see stock. |
||||||
10694491
|
BRAND™
704343 |
For bottle thread S 40 |
14.60 €
Each |
In Stock
Log in to see stock. |
||||||
10636161
|
BRAND™
704348 |
For bottle thread S 60 |
42.75 €
Each |
In Stock
Log in to see stock. |
||||||
10592423
|
BRAND™
704430 |
For bottle thread S 54 |
36.20 €
Each |
In Stock
Log in to see stock. |
||||||
Description
- Thread adapter, Polypropylene (PP), outer thread GL 45, for bottle thread S 40
- Thread Adapters for BRAND™ Bottle Top Dispensers, Polypropylene, 45/40 mm
- For use with Bottle Thread 40 S, Dispensette, Seripettor, Titrette and QuikSip Bottletop Diluters and Dispensers
- The Polypropylene (PP) and Ethylene Tetrafluoroethylene (ETFE) adapters are for general purpose use
- Ideal for use with Dispensetter bottletop dispensers
- These polypropylene and fluoropolymer (ETFE and PTFE) adapters are for use with BRAND™ bottletop dispensers and burettes
- These thread adapters help to maximize the compatibility of bottletop dispensers with most reagent bottles
Specifications
Thread Adapter | |
Polypropylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Polypropylene Thread Adapters