missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Polypropylene Thread Adapter
Buerkle™ Polypropylene Thread Adapter facilitates the secure connection of pumps and discharge cocks to a variety of containers.
Brand: Buerkle™ 0600-1006
22.63 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Constructed of polypropylene
- Interchange the connections to different adapters
Specifications
Thread Adapter | |
Pumps | |
External Thread Tri-Sure, Internal Thread 2 in. BSP |
Polypropylene | |
External: Tri-Sure; Internal: 2 in. (BSP) | |
Orange |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction