missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Macro-Line Pipette Tips
Eliminate sample contamination and prevent sample loss. Fisherbrand™ Macro-Line Pipette Tips provide precision pipetting with conical tip and secure fit.
29.54 €
Specifications
Volume (Metric) | 1 to 5 mL |
---|---|
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Length (Metric) | 126mm |
Certifications/Compliance | ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600 |
Product Code | Brand | Volume (Metric) | Sterility | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Color | Price | Quantity & Availability | ||||||
15285163
|
Fisherbrand™
FI1-210-84-0 |
1 to 5 mL | Non-sterile | Blue | N/A | N/A | N/A | |||||
Description
- Optimized cone geometry for a secure fit
- Graduation marks on tip at standard volumes
- Conical tip opening prevents drop formation at the end of the tip
- Tips made of autoclavable, non-wettable, virgin polypropylene
- Sterile packs available (beta irradiation)
Specifications
1 to 5 mL | |
Autoclavable | |
ISO Standard 8655, EU-Directive 98/79/EG, competent authority code DE/CA90, EN 552/ISO 11137 and DIN EN 556, EU Directive 2002/95/EC, ISO Standard 14000 and ISO Standard 14001, ISO Standard 10993, DIN Standard 12600 | |
Bulk | |
Universal Fit | |
Macro-Line |
Non-sterile | |
126mm | |
Autoclave | |
Universal | |
No | |
Virgin Polypropylene |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Macro-Line Pipette Tips