Learn More
BRAND™ 96-well Rack with Tubes
Well-suited for use with robots and other automated dispensing systems. Tubes, racks, and lids can withstand temperatures as low as –80°C. *Cover mat and strips are not autoclavable.
Brand:  BRAND™ 781562
 View more versions of this product
Product Code. 10093933
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Tubes and rack are autoclavable at 121°C (2 bar), acc. DIN EN 285 (cover mat/strip, TPE, not autoclavable). Tubes, racks and lids can withstand temperatures as low as -80°C. The seal of the tubes is pressure tested and there is a stable connection between the mounting plate and the racks, making these stackable racks well suited for use with robots and automated systems. Coded tubes (A1-H12) and rack sides are suitable for barcode labels.
Ideal for use with robots and other automated dispensing systems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN EN 285
Tube Rack
Autoclavable
Polypropylene
50 Pack
With Lid, no tubes
Yes
96
0.65mL Tubes