Learn More
Ratiolab™ Polypropylene Urine Beakers
Collect urine specimens in these beakers. Ratiolab™ Polypropylene Urine Beakers are ideal for sampling, transport and storage of solid, pasty and liquid samples.
Brand:  Ratiolab™ 5921175
 View more versions of this product
Product Code. 15187073
  • 83.30 € / Pack of 600
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in polypropylene (PP) or polystyrene (PS)
  • Practical multi-purpose beakers and bottles
  • Ideal for the clinical routine
  • With red screw-cap
  • Supplied in non-sterile or sterile, individually packed
Specifications
Specifications
600 Pk.
Urine Beaker
For sampling, transport and storage of solid, pasty and liquid samples
60 mL
Polypropylene
For sampling, transport and storage of solid, pasty and liquid samples