Learn More
Cole-Parmer™ Essentials Anti-Static Weighing Funnels, Polypropylene
Polypropylene Anti-Static Weighing Funnels ensure complete sample transfer.
Brand: Cole-Parmer™ Essentials 06127-04
200.30 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Autoclavable
- 6 month warranty
- These disposable polypropylene (PP) weighing funnels are designed to fit in volumetric flasks
- One side of the funnel is flattened so you can set the funnel flat on a balance
Specifications
Polypropylene | |
Size: Medium | |
Designed to Fit Microwell Plates |
Autoclavable | |
Volumetric flask | |
Weighing Funnel |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Cole-Parmer™ Essentials Anti-Static Weighing Funnels, Polypropylene > Anti-Static Weighing Funnels; Size: Medium
Spot an opportunity for improvement?Share a Content Correction