missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Thermo Scientific Chemicals

Product Code. 10698313
Click to view available options
Quantity:
5 mL
250 mL
1 L
5 L
25 L
Packaging Type:
Glass Bottle
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-64-5
Molecular Formula C26H50O10
Synonym Polyoxyethylene(20)sorbitan monolaurate

Specifications

Color Colorless to Yellow
Density 1.1000 g/mL
Packaging Type Plastic drum
Quantity 25 L
Solubility Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol
Form Liquid
Chemical Name or Material Polysorbate 20

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'