missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene, atactic, Thermo Scientific Chemicals

Product Code. 11415290
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Polystyrene, atactic is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-53-6
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Physical Form Beads
Solubility Information Insoluble in water.
Quantity 50 g
IUPAC Name styrene
Formula Weight 125,000-250,000
Odor Odorless
Chemical Name or Material Polystyrene, Atactic

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'