missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Snap Cap Vials

These vials are excellent for storing dry materials, powders, tablets, and other samples

Brand:  Bel-Art™ SP Scienceware™ F17575-0040

Product Code. 15371381

  • 137.56 € / Pack of 72
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Heavy, rigid-wall construction ensures long service
  • Can be used at temperatures from -20 to 70°C (-4 to 158°F)
  • Supplied with tight-fitting, polyethylene snap-on caps

Order Info

dimCapacityMetric

Specifications

Specifications

72 Pk.
Clear
Polystyrene
98 mm
148 mL
Container
Storing dry materials, Powders, Tablets
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers