missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Polystyrene Microscope Slide Box
Manufactured from virtually unbreakable high impact polystyrene. Kartell™ Polystyrene Microscope Slide Boxes are designed for meticulous storage and classification of microscope slides.
Brand: Kartell™ 0027800
13.86 EUR valid until 2024-09-30
Use promo code "21790" to get your promotional price.
This item is not returnable.
View return policy
Description
- Slots are well spaced for easy handling of 76mm x 26mm slides
- Slots are clearly numbered and can be cross referenced with corresponding index card
- Lids are totally removable
- Top and back clearly indicated
- The 50 and 100 place boxes have locating lugs so boxes can be stacked
Specifications
100 | |
100 | |
100 Slides | |
Lid and index card | |
Yes |
Gray | |
180 mm | |
35 mm | |
Polystyrene | |
230 mm |