missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Sterile Petri Dishes

These polystyrene Sterile Petri Dishes are disposable and have convenient stacking ridges.

Brand:  Cole-Parmer™ Essentials 06139-00

193.92 EUR valid until 2024-12-31
Use promo code "21955" to get your promotional price.


 View more versions of this product

Product Code. 15236464

  • 229.00 € / Pack of 500
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gamma irradiated.
  • Disposable.
  • With lid.
  • Convenient stacking ridges.
  • Packed sterile in easy-open bags of 20.
    • 2 boxes/case.
Specifications

Specifications

Single-use
Round
Petri Dish
500/Cs.
100 x 15 mm (3.9 x 0.6 in.)
0.6 in.
100 mm
Polystyrene
Sterile
For culturing fungi, bacteria, and other microorganisms
Bags
15 mm
3.9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.