missing translation for 'onlineSavingsMsg'
Learn More

Sterilin™ Polystyrene Petri Dishes
GREENER_CHOICE

Brand:  Sterilin™ 501V/IRR

170.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 12497612

  • 254.00 € / Pack of 80
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polystyrene
Petri Dish
80 Case
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sterilin™ Polystyrene Petri Dishes > Outer Diameter: 140mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.