missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polystyrene Serological Pipets

Brand:  Ratiolab™ 3110011

 View more versions of this product

Product Code. 10150843

  • 63.45 € / Pack of 200
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Glass Polystyrene
Yes
3 mL
Plastic/Paper
10 mL
Serological Pipet
Sterile
0.1 mL
200/Pk.
Clear
295 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers