Learn More
Fisherbrand™ Polystyrene Diamond-Shaped Standard Weighing Boat
Designed for accurate pouring with minimal sample loss. Fisherbrand™ Polystyrene Diamond-Shaped Standard Weighing Boat features a smooth surface that flexes easily. Disposable boats have a stable, flat bottom and can be used in a variety of lab environments and applications.
Brand:  Fisherbrand™ HS1427BF
 View more versions of this product
Product Code. 12962850
  • 56.85 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Features a diamond shape that easily flexes for accurate pouring
  • Wide, flat bottom helps resist tipping
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
  • Temperature range -10 to 70°C
  • Can be used with microwave moisture analyzers
  • 500 FDA/food-grade compliant boats per pack
Specifications
Specifications
Flat
Black
Polystyrene
Weighing Boat
78 x 56 x 14 mm
56 mm
30 mL
Single-use
Diamond
Microwave Moisture Analyzer
78 mm
14 mm