Learn More
Cole-Parmer™ CPC™ Plastic Quick-Disconnect Sealing Plug
Easy thumb-latch disconnection
Brand: Cole-Parmer™ 31311-78
This item is not returnable.
View return policy
Description
Simply Press the thumb latch to disconnect coupling halves.Specifications
Sealing Plug | |
-40°C | |
148°C |
Polysulfone | |
5 Pack |